Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001253719.1 | NP_001240648.1 | 6 | 813 | |
NM_178673.4 | NP_848788.2 | 6 | 1073 | ||
GenBank mRNA | AF374460.1 | - | 6 | 1047 | |
AK033652.1 | - | 6 | 1064 | ||
AK046070.1 | - | 6 | 814 | ||
AK081152.1 | - | 6 | 1064 | ||
AK138388.1 | - | 6 | 1365 | ||
AK140611.1 | - | 6 | 1076 | ||
BC075640.1 | - | 6 | 1036 | 30545529 |