Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_170593.3 | NP_733481.1 | 8 | 1437 | |
GenBank mRNA | AK035975.1 | - | 5 | 2126 | |
AK038938.1 | - | 7 | 2885 | ||
AK122547.1 | - | 1 | 66 | ||
AY150699.1 | - | 8 | 1238 | ||
BC118526.1 | - | 8 | 1232 | 40099214 |