Gene Transcripts

Gene Symbol
Fnip1
Entrez Gene ID
216742
Gene Name
folliculin interacting protein 1
Gene Aliases
A730024A03Rik, AI838773, AW557298
Chromosome Location
Chr. 11: 54438146 - 54518241 on Build GRCm38
UniGene ID
Mm.34087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173753.4 NP_776114.2 14 2665
GenBank mRNA AK042779.1 - 1 129
AK122574.1 - 14 2613
BC079892.1 - 14 2655 30543296

View Full Product Details