Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001081247.1 | NP_001074716.1 | 30 | 4118 | |
GenBank mRNA | AK170475.1 | - | 30 | 4132 | |
BC053071.1 | - | 29 | 4281 | ||
BC151002.1 | - | 30 | 4028 | 9087915 | |
BC151168.1 | - | 30 | 4026 | 9088057 | |
BC151169.1 | - | 30 | 4028 | 9088058 |