Gene Transcripts

Gene Symbol
Wapal
Entrez Gene ID
218914
Gene Name
wings apart-like homolog (Drosophila)
Gene Aliases
A530089A20Rik, BC037674, DIF-2, FOE, WAPL
Chromosome Location
Chr. 14: 34671487 - 34747983 on Build GRCm38
UniGene ID
Mm.27957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004436.4 NP_001004436.2 15 3517
NM_001301330.1 NP_001288259.1 15 3499
GenBank mRNA AB167349.1 - 15 3323
AK028167.1 - 13 1621
AK129105.1 - 15 3517
BC037674.1 - 9 1064
BC139434.1 - 15 3272 8862456
BC145625.1 - 15 3254 9054193

View Full Product Details