Gene Transcripts
- Gene Symbol
- Traf2
- Entrez Gene ID
- 22030
- Gene Name
- TNF receptor-associated factor 2
- Gene Aliases
- AI325259
- Chromosome Location
- Chr. 2: 25517982 - 25546940 on Build GRCm38
- UniGene ID
- Mm.3399
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001290413.1 |
NP_001277342.1
|
8, 9 |
1088 |
|
NM_009422.3 |
NP_033448.2
|
8, 9 |
1067 |
|
GenBank mRNA |
AF027570.1 |
- |
7, 8 |
979 |
|
AK038136.1 |
- |
8, 9 |
1016 |
|
AK052934.1 |
- |
8, 9 |
1016 |
|
AK151653.1 |
- |
8, 9 |
1005 |
|
AK152174.1 |
- |
8, 9 |
1035 |
|
AK159581.1 |
- |
8, 9 |
1037 |
|
AK159585.1 |
- |
|
1014 |
|
BC003801.1 |
- |
7, 8 |
993 |
3492839 |
BC060625.1 |
- |
6, 7 |
3024 |
|
L35303.1 |
- |
8, 9 |
1004 |
|
View Full Product Details