Gene Transcripts

Gene Symbol
Fhdc1
Entrez Gene ID
229474
Gene Name
FH2 domain containing 1
Gene Aliases
6330505N24Rik, Gm126
Chromosome Location
Chr. 3: 84442191 - 84480439 on Build GRCm38
UniGene ID
Mm.331107
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033301.4 NP_001028473.2 12 2506
NM_001205355.1 NP_001192284.1 12 2577
GenBank mRNA AK031946.1 - 11 1979
AK145217.1 - 12 2497
AK173244.1 - 12 2577
BC137632.1 - 12 2546 8860651

View Full Product Details