Gene Transcripts
- Gene Symbol
- Trmt13
- Entrez Gene ID
- 229780
- Gene Name
- tRNA methyltransferase 13
- Gene Aliases
- 4631408H19Rik, A330067P21, A930028L21Rik, Ccdc76
- Chromosome Location
- Chr. 3: 116581093 - 116614587 on Build GRCm38
- UniGene ID
- Mm.259648
- Species
- Mus musculus
- Gene Symbol
- Lrrc39
- Entrez Gene ID
- 109245
- Gene Name
- leucine rich repeat containing 39
- Gene Aliases
- 2010005E21Rik, 9430028I06Rik
- Chromosome Location
- Chr. 3: 116562953 - 116583134 on Build GRCm38
- UniGene ID
- Mm.216027
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC052542.1 |
- |
|
1089 |
|
View Full Product Details