Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145958.1 | NP_001139430.1 | 19 | 2695 | |
NM_172122.2 | NP_742120.2 | 20 | 2948 | ||
GenBank mRNA | AF527975.1 | - | 20 | 2948 | |
AK122285.1 | - | 12 | 1698 | ||
BC036729.1 | - | 5 | 686 | ||
BC054054.1 | - | 18 | 2676 | 6394697 | |
CA749493.1 | - | 5 | 619 |