Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001048192.2 | NP_001041657.1 | 8 | 1202 | |
NM_001294262.1 | NP_001281191.1 | 7 | 1230 | ||
NM_174849.3 | NP_777274.1 | 7 | 1119 | ||
GenBank mRNA | AK029676.1 | - | 7 | 1120 | |
AK034894.1 | - | 7 | 1208 | ||
AK035680.1 | - | 5 | 687 | ||
AK044195.1 | - | 6 | 2524 | ||
AK083656.1 | - | 7 | 1069 | ||
BC057349.1 | - | 7 | 1120 | 6813901 | |
DQ867034.1 | - | 7 | 1115 | ||
DQ867035.1 | - | 7 | 1115 | ||
DQ867036.1 | - | 8 | 1202 | ||
DQ867037.1 | - | 7 | 1115 | ||
DQ867038.1 | - | 7 | 1115 |