Gene Transcripts

Gene Symbol
Sdk2
Entrez Gene ID
237979
Gene Name
sidekick homolog 2 (chicken)
Gene Aliases
4632412F08Rik, 5330435L01Rik, Sdk-2, mKIAA1514
Chromosome Location
Chr. 11: 113776374 - 114066181 on Build GRCm38
UniGene ID
Mm.130504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172800.2 NP_766388.2 6, 7 820
GenBank mRNA AK030591.1 - 1918
AK052040.1 - 6, 7 845
AK129379.1 - 2, 3 201
AY351699.1 - 6, 7 820
BC038036.1 - 6, 7 1098

View Full Product Details