Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_175532.3 | NP_780741.1 | 3 | 418 | |
GenBank mRNA | AK028575.1 | - | 3 | 441 | |
AK028802.1 | - | 3 | 420 | ||
AK032446.1 | - | 3 | 414 | ||
AY355344.1 | - | 2 | 387 | ||
AY489193.1 | - | 2 | 371 | ||
BC113194.1 | - | 3 | 414 | 40062457 | |
BC113801.1 | - | 3 | 414 | 40062458 |