Gene Transcripts

Gene Symbol
Mrgpre
Entrez Gene ID
244238
Gene Name
MAS-related GPR, member E
Gene Aliases
C130069N09Rik, Ebrt3, Mgrf, MrgE
Chromosome Location
Chr. 7: 143778363 - 143784500 on Build GRCm38
UniGene ID
Mm.183561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175534.3 NP_780743.1 2 527
GenBank mRNA AJ320231.1 - 2 238
AK048528.1 - 2 345
AK133657.1 - 2 529
AK154886.1 - 2 322
BC096766.1 - 2 323 30102203
BC138305.1 - 2 236 8861325
BC138308.1 - 2 236 8861328

View Full Product Details