Gene Transcripts

Gene Symbol
Idh2
Entrez Gene ID
269951
Gene Name
isocitrate dehydrogenase 2 (NADP+), mitochondrial
Gene Aliases
E430004F23, IDPm, Idh-2
Chromosome Location
Chr. 7: 80094846 - 80115387 on Build GRCm38
UniGene ID
Mm.246432
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173011.2 NP_766599.2 2 233
GenBank mRNA AF212319.1 - 2 204
AK088110.1 - 2 205
AK145753.1 - 2 178
AK146564.1 - 2 216
AK161640.1 - 2 217
AK169395.1 - 2 230
BC060030.1 - 2 221 6515657
BY311397.1 - 2 242

View Full Product Details