Gene Transcripts

Gene Symbol
Zfp335
Entrez Gene ID
329559
Gene Name
zinc finger protein 335
Gene Aliases
1810045J01Rik, NIF-1, Nif1, Znf335
Chromosome Location
Chr. 2: 164891882 - 164911750 on Build GRCm38
UniGene ID
Mm.330776
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199027.2 NP_950192.2 5 814
GenBank mRNA AK157577.1 - 5 642
BC062921.1 - 5 814
BC151013.1 - 4 551 9087926
BC157954.1 - 4 551 9088127
BC172135.1 - 4 551

View Full Product Details