Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164195.1 | NP_001157667.1 | 11 | 1198 | |
NM_019961.3 | NP_064345.1 | 11 | 1198 | ||
GenBank mRNA | AF152996.1 | - | 11 | 1121 | |
AK005315.1 | - | 11 | 1054 | ||
AK005978.1 | - | 10 | 895 | ||
AK012827.1 | - | 11 | 1176 | ||
AK075820.1 | - | 11 | 1113 | ||
AK146934.1 | - | 11 | 1177 | ||
BC033415.1 | - | 11 | 1080 | 5008068 | |
BC037606.1 | - | 11 | 1161 | 5361347 |