Gene Transcripts

Gene Symbol
Ms4a4c
Entrez Gene ID
64380
Gene Name
membrane-spanning 4-domains, subfamily A, member 4C
Gene Aliases
5830413L19Rik, AI449185, Ms4a9
Chromosome Location
Chr. 19: 11404717 - 11427246 on Build GRCm38
UniGene ID
Mm.440715
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029499.3 NP_083775.1 3 359
GenBank mRNA AF237914.2 - 3 333
AK017928.1 - 3 339
AK151045.1 - 3 352
AK153495.1 - 2 203
AK157137.1 - 361
AK172650.1 - 3 331
BC027496.1 - 3 315 1383416
CT010170.1 - 2 227

View Full Product Details