Gene Transcripts

Gene Symbol
Crls1
Entrez Gene ID
66586
Gene Name
cardiolipin synthase 1
Gene Aliases
0610009I22Rik, 4930557M15Rik, 5730490M08Rik
Chromosome Location
Chr. 2: 132846650 - 132866786 on Build GRCm38
UniGene ID
Mm.357342
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024385.1 NP_001019556.1 2 529
NM_025646.3 NP_079922.2 2 216
GenBank mRNA AK002408.1 - 2 531
AK004215.1 - 2 245
AK016165.1 - 2 217
AK017718.1 - 2 508
AK018180.1 - 2 530
BC048702.1 - 2 510 6506241
BC086619.1 - 2 488

View Full Product Details