Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024385.1 | NP_001019556.1 | 3, 4 | 735 | |
NM_025646.3 | NP_079922.2 | 3, 4 | 422 | ||
GenBank mRNA | AK002408.1 | - | 3, 4 | 737 | |
AK004215.1 | - | 3, 4 | 451 | ||
AK016165.1 | - | 3, 4 | 423 | ||
BC048702.1 | - | 3, 4 | 716 | 6506241 | |
BC086619.1 | - | 3, 4 | 692 |