Gene Transcripts

Gene Symbol
Saraf
Entrez Gene ID
67887
Gene Name
store-operated calcium entry-associated regulatory factor
Gene Aliases
1810045K07Rik, Tmem66
Chromosome Location
Chr. 8: 34154563 - 34170847 on Build GRCm38
UniGene ID
Mm.290353
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026432.3 NP_080708.3 1, 2 273
GenBank mRNA AK007787.1 - 1, 2 200
AK075744.1 - 1, 2 164
AK077118.1 - 1, 2 261
AK088728.1 - 1, 2 156
AK093942.1 - 1, 2 248
AK163218.1 - 1, 2 206
BC013497.1 - 1, 2 214 4222385
BC022616.1 - 1, 2 251 4187981
BC024888.1 - 1, 2 209 3988822
BY135425.1 - 1, 2 273

View Full Product Details