Gene Transcripts

Gene Symbol
Mphosph10
Entrez Gene ID
67973
Gene Name
M-phase phosphoprotein 10 (U3 small nucleolar ribonucleoprotein)
Gene Aliases
2810453H10Rik, 5730405D16Rik, AI326008
Chromosome Location
Chr. 7: 64376541 - 64392236 on Build GRCm38
UniGene ID
Mm.26973
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026483.2 NP_080759.2 10 1893
GenBank mRNA AK008785.1 - 7 823
AV222458.2 - 3 384
BC029198.1 - 2 282

View Full Product Details