Gene Transcripts

Gene Symbol
Eml1
Entrez Gene ID
68519
Gene Name
echinoderm microtubule associated protein like 1
Gene Aliases
1110008N23Rik, A930030P13Rik, AA171013, AI847476, AI853955, ELP79, EMAP, EMAPL
Chromosome Location
Chr. 12: 108371153 - 108539564 on Build GRCm38
UniGene ID
Mm.236645
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001043335.1 NP_001036800.1 10 1447
NM_001043336.2 NP_001036801.1 10 1064
NM_001286346.1 NP_001273275.1 10 1201
NM_001286347.1 NP_001273276.1 11 1115
GenBank mRNA AK044662.1 - 10 1186
BC053094.1 - 10 1281 6400458
BC059839.1 - 10 1042 6417403
BC079582.1 - 10 1100 5702743
BC098329.1 - 9 844
BC125289.1 - 11 1086 40129804
BC144757.1 - 11 1086
BC150975.1 - 10 1201 9087888

View Full Product Details