Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027000.4 | NP_081276.2 | 13 | 1325 | |
GenBank mRNA | AF325354.1 | - | 13 | 1309 | |
AF348208.1 | - | 13 | 1332 | ||
AK032309.1 | - | 1 | 20 | ||
AK141852.1 | - | 13 | 1335 | ||
AK163372.1 | - | 12 | 1145 | ||
AK164330.1 | - | 13 | 1326 | ||
AK166711.1 | - | 13 | 1332 | ||
BC005514.1 | - | 13 | 1306 | ||
BC025431.1 | - | 13 | 1330 | 5007174 |