Gene Transcripts

Gene Symbol
Rhobtb1
Entrez Gene ID
69288
Gene Name
Rho-related BTB domain containing 1
Gene Aliases
1700008H16Rik, 3110048G13Rik, AI173445, AI573858, AV350930
Chromosome Location
Chr. 10: 69151279 - 69291791 on Build GRCm38
UniGene ID
Mm.26659
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081347.1 NP_001074816.1 8 1741
NM_001252636.1 NP_001239565.1 8 1724
NM_001252637.1 NP_001239566.1 8 1845
NM_001252638.1 NP_001239567.1 9 1904
GenBank mRNA AK005770.1 - 8 1724
AK078306.1 - 7 1678
AK173015.1 - 3 1105
BC099973.1 - 8 1722 6432524
BC137646.1 - 7 1675 8860665
BC137647.1 - 7 1675 8860666
BC144800.1 - 8 1734 9053348

View Full Product Details