Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027444.3 | NP_081720.2 | 12 | 2332 | |
GenBank mRNA | AF454943.1 | - | 12 | 2325 | |
AF454944.1 | - | 10 | 2197 | ||
AK029532.1 | - | 10 | 2199 | ||
AK029747.1 | - | 13 | 2446 | ||
AK049516.1 | - | 12 | 2318 | ||
AK167724.1 | - | 12 | 2463 | ||
BC066821.1 | - | 10 | 1959 | 30061364 |