Gene Transcripts

Gene Symbol
Lrwd1
Entrez Gene ID
71735
Gene Name
leucine-rich repeats and WD repeat domain containing 1
Gene Aliases
1200011O22Rik, AU042569, AW548074, Orca
Chromosome Location
Chr. 5: 136123066 - 136136074 on Build GRCm38
UniGene ID
Mm.425292
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027891.4 NP_082167.2 8 1091
GenBank mRNA AK004717.1 - 6 2053
AK051910.1 - 8 1039
AK084948.1 - 8 1091
AK153976.1 - 8 1062
AK154352.1 - 8 1033
AK167020.1 - 8 1040
AK172708.1 - 8 1059
BC048397.1 - 8 1070 6396815

View Full Product Details