Gene Transcripts

Gene Symbol
Zswim2
Entrez Gene ID
71861
Gene Name
zinc finger SWIM-type containing 2
Gene Aliases
1700025P14Rik, 4933437F18Rik, MEX
Chromosome Location
Chr. 2: 83915079 - 83941254 on Build GRCm38
UniGene ID
Mm.116957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027964.2 NP_082240.1 8, 9 1145
GenBank mRNA AK006367.1 - 8, 9 1146
BC120837.1 - 8, 9 1125 40129760
EU233996.1 - 6, 7 1260

View Full Product Details