Gene Transcripts

Gene Symbol
Mfsd6
Entrez Gene ID
98682
Gene Name
major facilitator superfamily domain containing 6
Gene Aliases
2210010L05Rik, 9630025I22Rik, AI836200, AW212394, MMR2
Chromosome Location
Chr. 1: 52656299 - 52727462 on Build GRCm38
UniGene ID
Mm.491173
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133829.2 NP_598590.2 3 1914
NM_178081.4 NP_835182.1 3 1914
GenBank mRNA AB247936.1 - 2 1529
AK035995.1 - 3 1915
AK046866.1 - 3 2058
AK171946.1 - 3 1955
BC017534.1 - 2 1023 4488390
BC029324.1 - 2 1271

View Full Product Details