Gene Transcripts

Gene Symbol
Rab20
Entrez Gene ID
19332
Gene Name
RAB20, member RAS oncogene family
Gene Aliases
AA536966, D8Ertd350e
Chromosome Location
Chr. 8: 11453513 - 11478499 on Build GRCm38
UniGene ID
Mm.37632
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011227.1 NP_035357.1 2 688
GenBank mRNA AK152454.1 - 2 814
AK163743.1 - 2 828
AK164541.1 - 2 828
BG066967.2 - 1 486

View Full Product Details