Gene Transcripts

Gene Symbol
Krt73
Entrez Gene ID
223915
Gene Name
keratin 73
Gene Aliases
BC067067, Kb36
Chromosome Location
Chr. 15: 101793308 - 101802332 on Build GRCm38
UniGene ID
Mm.291498
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_212485.2 NP_997650.1 9 1567
GenBank mRNA BC067067.1 - 9 1567

View Full Product Details