Gene Transcripts

Gene Symbol
Tfap2e
Entrez Gene ID
332937
Gene Name
transcription factor AP-2, epsilon
Gene Aliases
AI505200, AP-2e, Tcfap2e
Chromosome Location
Chr. 4: 126716003 - 126736269 on Build GRCm38
UniGene ID
Mm.331881
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198960.2 NP_945198.2 7 1421
GenBank mRNA AY325902.1 - 7 1261
DV649306.1 - 2 225

View Full Product Details