Gene Transcripts

Gene Symbol
Maml2
Entrez Gene ID
270118
Gene Name
mastermind like 2 (Drosophila)
Gene Aliases
5930431H10, BC032967
Chromosome Location
Chr. 9: 13297958 - 13298561 on Build GRCm38
UniGene ID
Mm.116898
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013813.3 NP_001013835.2 752
GenBank mRNA BC053388.1 - 764

View Full Product Details