Gene Transcripts

Gene Symbol
Nradd
Entrez Gene ID
67169
Gene Name
neurotrophin receptor associated death domain
Gene Aliases
2610311B09Rik, BB104149, NRDD, NRH2, PLAIDD
Chromosome Location
Chr. 9: 110621131 - 110624393 on Build GRCm38
UniGene ID
Mm.46680
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026012.2 NP_080288.1 4 537
GenBank mRNA AF534394.1 - 3 349
AK007483.1 - 2 194
AK160222.1 - 4 523
BC039808.1 - 4 507 1547029
BC069856.1 - 4 537 6491606

View Full Product Details