Gene Transcripts
- Gene Symbol
- Fbxl2
- Entrez Gene ID
- 72179
- Gene Name
- F-box and leucine-rich repeat protein 2
- Gene Aliases
- 2810423A21Rik, Fbl3
- Chromosome Location
- Chr. 9: 113958757 - 114026787 on Build GRCm38
- UniGene ID
- Mm.386808
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_178624.6 |
NP_848739.1
|
5, 6 |
342 |
|
GenBank mRNA |
AK013142.1 |
- |
6, 7 |
476 |
|
AK039010.1 |
- |
5, 6 |
342 |
|
AK044693.1 |
- |
5, 6 |
339 |
|
AK045742.1 |
- |
5, 6 |
329 |
|
AK089994.1 |
- |
5, 6 |
318 |
|
AK141177.1 |
- |
5, 6 |
332 |
|
AK163472.1 |
- |
5, 6 |
338 |
|
BC096582.1 |
- |
5, 6 |
328 |
3963599 |
BC145665.1 |
- |
5, 6 |
322 |
40130968 |
BC145998.1 |
- |
5, 6 |
322 |
40131301 |
View Full Product Details