Gene Transcripts

Gene Symbol
Pcdhb7
Entrez Gene ID
93878
Gene Name
protocadherin beta 7
Gene Aliases
Pcdhb4B, PcdhbG
Chromosome Location
Chr. 18: 37341702 - 37345207 on Build GRCm38
UniGene ID
Mm.44755
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053132.3 NP_444362.3 1 658
GenBank mRNA AF326300.1 - 1 454
AK029340.1 - 1 637
AK034205.1 - 1 660
AK043979.1 - 1 617
AK046463.1 - 1 641
AY013789.1 - 1 454
BC094897.1 - 1 620

View Full Product Details