Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023239.4 | NP_075728.1 | 1 | 704 | |
GenBank mRNA | AF319979.1 | - | 1 | 582 | |
AK010294.1 | - | 1 | 665 | ||
AK017727.1 | - | 1 | 706 | ||
AK018250.1 | - | 1 | 665 | ||
AK049759.1 | - | 1 | 684 | ||
AK076471.1 | - | 1 | 690 | ||
AK131157.1 | - | 661 | |||
BC034892.1 | - | 1 | 629 | 1494952 | |
BC092289.1 | - | 1 | 666 | 4990134 |