Gene Transcripts

Gene Symbol
Aldh3b1
Entrez Gene ID
67689
Gene Name
aldehyde dehydrogenase 3 family, member B1
Gene Aliases
1700001N19Rik, ALDH4, ALDH7
Chromosome Location
Chr. 19: 3913491 - 3930817 on Build GRCm38
UniGene ID
Mm.109341
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026316.2 NP_080592.2 3 266
GenBank mRNA AF362571.1 - 2 188
AK154500.1 - 3 312
AK154723.1 - 3 359
AK159447.1 - 3 280
AK160106.1 - 3 283
BC023305.1 - 3 287
BC046597.1 - 3 266 6312211

View Full Product Details