Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001164604.1 | NP_001158076.1 | 3 | 422 | |
NM_026746.3 | NP_081022.2 | 3 | 422 | ||
GenBank mRNA | AK003685.1 | - | 3 | 374 | |
AK051277.1 | - | 3 | 374 | ||
AK211545.1 | - | 3 | 303 | ||
AK212165.1 | - | 3 | 303 | ||
BC009125.1 | - | 3 | 333 | 3155917 | |
DV057668.1 | - | 3 | 442 |