Gene Transcripts

Gene Symbol
Meig1
Entrez Gene ID
104362
Gene Name
meiosis expressed gene 1
Gene Aliases
MLZ-278, Meg1
Chromosome Location
Chr. 2: 3409043 - 3422648 on Build GRCm38
UniGene ID
Mm.2688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008579.4 NP_032605.1 3 478
GenBank mRNA AK006347.1 - 3 292
AK007239.1 - 3 292
BC049538.1 - 3 478 6773052
X64455.1 - 3 288

View Full Product Details