Gene Transcripts

Gene Symbol
Pnrc1
Entrez Gene ID
108767
Gene Name
proline-rich nuclear receptor coactivator 1
Gene Aliases
5730463C12Rik, B4-2, PRR2, Prol2
Chromosome Location
Chr. 4: 33245423 - 33248787 on Build GRCm38
UniGene ID
Mm.27769
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033225.2 NP_001028397.2 2 1150
GenBank mRNA AK087995.1 - 2 875
AK159693.1 - 2 873
AK160284.1 - 2 874
BC038496.1 - 1 180

View Full Product Details