Gene Transcripts

Gene Symbol
Uhrf2
Entrez Gene ID
109113
Gene Name
ubiquitin-like, containing PHD and RING finger domains 2
Gene Aliases
2310065A22Rik, AI426270, AW214556, D130071B19Rik, Nirf
Chromosome Location
Chr. 19: 30030433 - 30093729 on Build GRCm38
UniGene ID
Mm.313364
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144873.2 NP_659122.2 9, 10 1777
GenBank mRNA AB116653.1 - 9, 10 1512
AF274047.1 - 9, 10 1687
BC060241.1 - 9, 10 1777 6334856

View Full Product Details