Gene Transcripts

Gene Symbol
Alox5
Entrez Gene ID
11689
Gene Name
arachidonate 5-lipoxygenase
Gene Aliases
5-LO, 5-LOX, 5LO, 5LX, AI850497, F730011J02
Chromosome Location
Chr. 6: 116410069 - 116461210 on Build GRCm38
UniGene ID
Mm.41072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009662.2 NP_033792.1 6 959
GenBank mRNA AK089344.1 - 6 913
AK136868.1 - 6 928
AK137481.1 - 6 917
AK169938.1 - 6 916
AK170016.1 - 6 816
AK170434.1 - 6 916
AK170879.1 - 6 915
AK171191.1 - 6 908
AK171413.1 - 6 916
BC139102.1 - 6 886 8862124
BC141213.1 - 6 886 9055825
L42198.1 - 6 805

View Full Product Details