Gene Transcripts

Gene Symbol
Amh
Entrez Gene ID
11705
Gene Name
anti-Mullerian hormone
Gene Aliases
MIS
Chromosome Location
Chr. 10: 80805021 - 80807679 on Build GRCm38
UniGene ID
Mm.376094
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007445.2 NP_031471.2 4 679
GenBank mRNA AY911505.1 - 4 679
BC150154.1 - 4 678
BC150477.1 - 4 678

View Full Product Details