Gene Transcripts

Gene Symbol
Bcl7c
Entrez Gene ID
12055
Gene Name
B cell CLL/lymphoma 7C
Gene Aliases
C230096E12Rik
Chromosome Location
Chr. 7: 127661454 - 127708780 on Build GRCm38
UniGene ID
Mm.89667
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009746.2 NP_033876.1 2 380
GenBank mRNA AK049064.1 - 2 400
AK131935.1 - 2 269
BC005673.1 - 2 380 3153237
BC058530.1 - 2 251 6818556
Y11905.1 - 3 198

View Full Product Details