Gene Transcripts

Gene Symbol
Clns1a
Entrez Gene ID
12729
Gene Name
chloride channel, nucleotide-sensitive, 1A
Gene Aliases
2610036D06Rik, 2610100O04Rik, Clci, Clcni, ICLN
Chromosome Location
Chr. 7: 97696657 - 97720793 on Build GRCm38
UniGene ID
Mm.21482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023671.2 NP_076160.1 5 710
GenBank mRNA AK011691.1 - 5 691
AK011789.1 - 5 696
AK031877.1 - 5 692
AK078050.1 - 5 663
AK133981.1 - 5 703
AK151356.1 - 5 695
AK151944.1 - 5 695
AK169259.1 - 5 674
BC005575.1 - 5 676 3967933
CX236388.1 - 5 709

View Full Product Details