Gene Transcripts

Gene Symbol
Endog
Entrez Gene ID
13804
Gene Name
endonuclease G
Gene Aliases
Chromosome Location
Chr. 2: 30171401 - 30174069 on Build GRCm38
UniGene ID
Mm.474473
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007931.1 NP_031957.1 1 453
GenBank mRNA AB012108.1 - 1 453
AK144459.1 - 1 485
BC030177.1 - 1 462 5029633

View Full Product Details