Gene Transcripts

Gene Symbol
Hipk1
Entrez Gene ID
15257
Gene Name
homeodomain interacting protein kinase 1
Gene Aliases
1110062K04Rik, Myak
Chromosome Location
Chr. 3: 103739815 - 103791894 on Build GRCm38
UniGene ID
Mm.20827
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001301304.1 NP_001288233.1 2 368
NM_001301306.1 NP_001288235.1 2 478
NM_010432.2 NP_034562.2 2 478
GenBank mRNA AF071070.1 - 2 368
AF071071.1 - 2 366
AF077658.1 - 2 382
AF170303.1 - 2 278
AF170304.1 - 2 278
AK134408.1 - 2 479
BC145697.1 - 2 231 40131000
BC145699.1 - 2 231 40131002
DQ279088.1 - 1 40

Gene Symbol
Gm15886
Entrez Gene ID
100379612
Gene Name
predicted gene 15886
Gene Aliases
OTTMUSG00000027335
Chromosome Location
Chr. : - on Build
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK016162.1 - 5 839

View Full Product Details