Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001130419.2 | NP_001123891.1 | 2 | 268 | |
NM_001286081.1 | NP_001273010.1 | 2 | 181 | ||
NM_001286083.1 | NP_001273012.1 | 2 | 113 | ||
NM_010471.4 | NP_034601.1 | 2 | 172 | ||
GenBank mRNA | AF326551.1 | - | 1 | 295 | |
AF326552.1 | - | 1 | 295 | ||
AK002992.1 | - | 2 | 172 | ||
BC049607.1 | - | 2 | 137 | 6595363 | |
BC058588.1 | - | 1 | 86 | 5360894 | |
BY129411.1 | - | 2 | 161 | ||
CX238650.1 | - | 2 | 145 | ||
CX239660.1 | - | 3 | 267 | ||
CX240088.1 | - | 2 | 113 | ||
CX240757.1 | - | 2 | 161 |