Gene Transcripts

Gene Symbol
Kap
Entrez Gene ID
16483
Gene Name
kidney androgen regulated protein
Gene Aliases
AW146435, FKSG22
Chromosome Location
Chr. 6: 133849855 - 133853667 on Build GRCm38
UniGene ID
Mm.13052
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010594.2 NP_034724.1 2 148
GenBank mRNA AK142506.1 - 2 149
AV000934.1 - 2 165
BC013453.1 - 2 134 4221610
M22810.1 - 2 147

View Full Product Details