Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136068.2 | NP_001129540.1 | 1 | 224 | |
NM_001288664.1 | NP_001275593.1 | 1 | 224 | ||
NM_010652.2 | NP_034782.1 | 1 | 224 | ||
GenBank mRNA | AF095447.1 | - | 1 | 161 | |
AF106008.1 | - | 1 | 187 | ||
AF106009.1 | - | 1 | 179 | ||
AF109782.1 | - | 1 | 161 | ||
AF109783.1 | - | 161 | |||
AF109784.1 | - | 1 | 161 | ||
AK138801.1 | - | 1 | 180 |